2-Chloro-α,α,α-trifluorotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-(trifluoromethyl)benzene, 2-Chlorobenzotrifluoride
PC1690-100G PC1690-1KG PC1690-500G PC1690-2.5KG PC1690-5KG PC1690-10KG
APOSPC1690-100GEA 21.9 EUR
APOSPC1690-100G APOSPC1690-1KG APOSPC1690-500G APOSPC1690-2.5KG APOSPC1690-5KG APOSPC1690-10KG
2-Chloro-α,α,α-trifluorotoluene 99%
2-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 152 °C
Melting Pt: –7 °C
Density: 1.379 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00000561
CAS Number: 88-16-4
UN: 2234
ADR: 3 III