2,6-Difluoro-3-nitrobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC1691-5G PC1691-1G PC1691-25G
APOSPC1691-5GEA 24.8 EUR
APOSPC1691-5G APOSPC1691-1G APOSPC1691-25G
2,6-Difluoro-3-nitrobenzoic acid 98%
2,6-Difluoro-3-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MW: 203.1 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00119291
CAS Number: 83141-10-0