2-Chloro-4-fluoro-5-nitrobenzoic acid 96%
Supplier: Apollo Scientific
Warning
PC1693-25G PC1693-5G PC1693-100G
APOSPC1693-25GEA 166 EUR
APOSPC1693-25G APOSPC1693-5G APOSPC1693-100G
2-Chloro-4-fluoro-5-nitrobenzoic acid 96%
2-Chloro-4-fluoro-5-nitrobenzoic acid
Formula: C₇H₃ClFNO
MW: 219.55 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD04115715
CAS Number: 114776-15-7