Potassium-2-(methylthio)phenyltrifluoroborate 96%
Supplier: Apollo Scientific
Synonyms: Potassium trifluoro(2-(methylthio)phenyl)borate, Potassium(2-Methylsulphonylphenyl)trifluoroboroate
PC1699-100G PC1699-25G PC1699-5G
APOSPC1699-100GEA 786 EUR
APOSPC1699-100G APOSPC1699-25G APOSPC1699-5G
Potassium-2-(methylthio)phenyltrifluoroborate 96%
Potassium 2-(methylthio)phenyltrifluoroborate
Formula: C₇H₇BF₃KS
MW: 230.10 g/mol
Melting Pt: 274…276 °C
Storage Temperature: Ambient
MDL Number: MFCD04115773
CAS Number: 850623-77-7