3-Chloro-α,α,α-trifluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-3-(trifluoromethyl)benzene, 3-Chlorobenzotrifluoride
PC1700-100G PC1700-250G
APOSPC1700-100GEA 40.6 EUR
APOSPC1700-100G APOSPC1700-250G
3-Chloro-α,α,α-trifluorotoluene 98%
3-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 137…138 °C
Melting Pt: –56 °C
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00000594
CAS Number: 98-15-7
EINECS: 202-642-9
UN: 2234
ADR: 3 III