2-Chloro-5-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-5-trifluoromethylbenzonitrile, 4-Chloro-3-cyanobenzotrifluoride
PC1714-100G PC1714-25G PC1714-5G
APOSPC1714-100GEA 112 EUR
APOSPC1714-100G APOSPC1714-25G APOSPC1714-5G
2-Chloro-5-(trifluoromethyl)benzonitrile 98%
2-Chloro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
Boiling Pt: 210…212 °C
Melting Pt: 36…42 °C
Density: 1.481 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00019742
CAS Number: 328-87-0
EINECS: 206-338-7
UN: 3439
ADR: 6.1,III