1-Chloro-2,6-dibromo-4-fluorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dibromo-2-chloro-5-fluorobenzene, 2-Chloro-1,3-dibromo-5-fluorobenzene
PC1717L-25G PC1717L-100G
APOSPC1717L-25GEA 34.8 EUR
APOSPC1717L-25G APOSPC1717L-100G
1-Chloro-2,6-dibromo-4-fluorobenzene
1-Chloro-2,6-dibromo-4-fluorobenzene
Formula: C₆H₂Br₂ClF
MW: 288.34 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00061122
CAS Number: 179897-90-6