5-Fluoro-2-nitrobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC1745-1G PC1745-25G PC1745-5G
APOSPC1745-1GEA 21.9 EUR
APOSPC1745-1G APOSPC1745-25G APOSPC1745-5G
5-Fluoro-2-nitrobenzonitrile 98%
5-Fluoro-2-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01632196
CAS Number: 50594-78-0