2-Fluoro-m-anisaldehyde 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-3-methoxybenzaldehyde, 2-Fluoro-3-anisaldehyde
PC1746-25G PC1746-5G PC1746-1G PC1746-10G
APOSPC1746-25GEA 209 EUR
APOSPC1746-25G APOSPC1746-5G APOSPC1746-1G APOSPC1746-10G
2-Fluoro-m-anisaldehyde 99%
2-Fluoro-m-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 114 °C/15mmHg
Melting Pt: 47…51 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03411962
CAS Number: 103438-88-6