Methyl-3-bromo-4-fluorobenzoate 98%
Supplier: Apollo Scientific
Warning
PC1748-25G PC1748-5G PC1748-100G
APOSPC1748-25GEA 101 EUR
APOSPC1748-25G APOSPC1748-5G APOSPC1748-100G
Methyl-3-bromo-4-fluorobenzoate 98%
Methyl 3-bromo-4-fluorobenzoate
Formula: C₈H₆BrFO₂
Melting Pt: 26…28 °C
Storage Temperature: Ambient
MDL Number: MFCD04972915
CAS Number: 82702-31-6