4-Chloro-1,3-difluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2,4-difluorobenzene
PC1754E-100G PC1754E-250G
APOSPC1754E-100GEA 52.2 EUR
APOSPC1754E-100G APOSPC1754E-250G
4-Chloro-1,3-difluorobenzene 98%
4-Chloro-1,3-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 127…128 °C
Melting Pt: –26 °C
Density: 1.353 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00042569
CAS Number: 1435-44-5
UN: 1993
ADR: 3,III