2-Chloro-1,4-difluorobenzene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,5-Difluorochlorobenzene
PC1754F-100G PC1754F-25G PC1754F-500G
APOSPC1754F-100GEA 65.4 EUR
APOSPC1754F-100G APOSPC1754F-25G APOSPC1754F-500G
2-Chloro-1,4-difluorobenzene 98%
2-Chloro-1,4-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 126…128 °C
Density: 1.352 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00042571
CAS Number: 2367-91-1
UN: 1993
ADR: 3,II