4-Chloro-1,2-difluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Difluorochlorobenzene
PC1754J-100G PC1754J-25G
APOSPC1754J-100GEA 119 EUR
APOSPC1754J-100G APOSPC1754J-25G
4-Chloro-1,2-difluorobenzene 98%
4-Chloro-1,2-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 126 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00042572
CAS Number: 696-02-6
UN: 1993
ADR: 3 III