2-Chloro-4,5-difluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC1756-100G PC1756-25G PC1756-500G
APOSPC1756-100GEA 185 EUR
APOSPC1756-100G APOSPC1756-25G APOSPC1756-500G
2-Chloro-4,5-difluorobenzoic acid 98%
2-Chloro-4,5-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Boiling Pt: 257…259 °C
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00077479
CAS Number: 110877-64-0
EINECS: 405-380-5