4-Fluoro-1-indanone 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-2,3-dihydro-1H-inden-1-one
PC1783-5G
APOSPC1783-5GEA 62.5 EUR
APOSPC1783-5G
4-Fluoro-1-indanone 95+%
4-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.152 g/mol
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00797829
CAS Number: 699-99-0