4-Bromo-2-(trifluoromethyl)phenol 99%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-hydroxybenzotrifluoride
PC1794-1G PC1794-5G PC1794-250MG
APOSPC1794-1GEA 29 EUR
APOSPC1794-1G APOSPC1794-5G APOSPC1794-250MG
4-Bromo-2-(trifluoromethyl)phenol 99%
4-Bromo-2-(trifluoromethyl)phenol
Formula: C₇H₄BrF₃O
MW: 241.01
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD05668981
CAS Number: 50824-04-9