Methyl-2-bromo-5-fluorobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-bromo-5-fluorobenzoate
PC1796-100G PC1796-25G PC1796-5G
APOSPC1796-100GEA 126 EUR
APOSPC1796-100G APOSPC1796-25G APOSPC1796-5G
Methyl-2-bromo-5-fluorobenzoate 98%
Methyl 2-bromo-5-fluorobenzoate
Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00086167
CAS Number: 6942-39-8