2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-1,5-dinitro-3-(trifluoromethyl)benzene, 2-Chloro-3,5-dinitrobenzotrifluoride, 1-Chloro-2,4-dinitro-6-(trifluoromethyl)benzene
PC1808-25G PC1808-5G PC1808-100G
APOSPC1808-25GEA 36.4 EUR
APOSPC1808-25G APOSPC1808-5G APOSPC1808-100G
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 99%
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Formula: C₇H₂ClF₃N₂O₄
MW: 270.55 g/mol
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00007076
CAS Number: 392-95-0
EINECS: 206-883-0