4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Chloro-2,6-dinitro-4-(trifluoromethyl)benzene, 4-Chloro-3,5-dinitrobenzotrifluoride
PC1810-100G PC1810-25G
APOSPC1810-100GEA 50.9 EUR
APOSPC1810-100G APOSPC1810-25G
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 97%
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Formula: C₇H₂ClF₃N₂O₄
MW: 270.55 g/mol
Boiling Pt: >250 °C
Melting Pt: 50…58 °C
Density: 1.6 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007068
CAS Number: 393-75-9
EINECS: 206-889-3
UN: 2811
ADR: 6.1 II