2,6-Dichloro-3-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC1811-1G PC1811-25G PC1811-5G
APOSPC1811-1GEA 27.7 EUR
APOSPC1811-1G APOSPC1811-25G APOSPC1811-5G
2,6-Dichloro-3-fluorobenzoic acid 98%
2,6-Dichloro-3-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658165
CAS Number: 178813-78-0