2'-Chloro-4'-fluoroacetophenone 98%
Supplier: Apollo Scientific
Warning
PC1817M-5G PC1817M-25G PC1817M-100G
APOSPC1817M-5GEA 21.9 EUR
APOSPC1817M-5G APOSPC1817M-25G APOSPC1817M-100G
2'-Chloro-4'-fluoroacetophenone 98%
2'-Chloro-4'-fluoroacetophenone
Formula: C₈H₆ClFO
Boiling Pt: 110 °C/19mmHg
Density: 1.3 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00042202
CAS Number: 700-35-6