3'-Chloro-4'-fluoroacetophenone 99%
Supplier: Apollo Scientific
Warning
PC1818M-100G PC1818M-25G
APOSPC1818M-100GEA 116 EUR
APOSPC1818M-100G APOSPC1818M-25G
3'-Chloro-4'-fluoroacetophenone 99%
3'-Chloro-4'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 126…128 °C/15mmHg
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00042203
CAS Number: 2923-66-2