2-Chloro-4-fluoroaniline
Supplier: Apollo Scientific
|
Warning
|
PC1819-25G
PC1819-100G
APOSPC1819-25GEA
21.9
EUR
APOSPC1819-25G
APOSPC1819-100G
2-Chloro-4-fluoroaniline
2-Chloro-4-fluoroaniline
Formula:
C₆H₅ClFN Boiling Pt: 191…192 °C Density: 1.219 g/cm³ (25 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042530 CAS Number: 2106-02-7 EINECS: 218-282-0 UN: 2810 ADR: 6.1,III |