3-Chloro-2-fluoroaniline 97%
Supplier: Apollo Scientific
Danger
PC1819W-25G PC1819W-100G PC1819W-500G
APOSPC1819W-25GEA 21.9 EUR
APOSPC1819W-25G APOSPC1819W-100G APOSPC1819W-500G
3-Chloro-2-fluoroaniline 97%
3-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 214 °C
Density: 1.324 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00069415
CAS Number: 2106-04-9
EINECS: 218-283-6
UN: 2810
ADR: 6.1,III