4-Chloro-2-fluoroaniline 98%
Supplier: Apollo Scientific
Danger
PC1822-100G PC1822-25G PC1822-250G PC1822-500G
APOSPC1822-100GEA 36.4 EUR
APOSPC1822-100G APOSPC1822-25G APOSPC1822-250G APOSPC1822-500G
4-Chloro-2-fluoroaniline 98%
4-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
Boiling Pt: 92 °C/2mmHg
Density: 1.311 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00010625
CAS Number: 57946-56-2
EINECS: 261-034-1
UN: 2810
ADR: 6.1,III