2-Bromo-5-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC1825-100G PC1825-500G
APOSPC1825-100GEA 59.6 EUR
APOSPC1825-100G APOSPC1825-500G
2-Bromo-5-fluorobenzoic acid 98%
2-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00142874
CAS Number: 394-28-5