1-Chloro-2-fluorobenzene 99%
Supplier: Apollo Scientific
Warning
PC1840-100G PC1840-25G PC1840-5G
APOSPC1840-100GEA 132 EUR
APOSPC1840-100G APOSPC1840-25G APOSPC1840-5G
1-Chloro-2-fluorobenzene 99%
1-Chloro-2-fluorobenzene
Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 137…138 °C
Melting Pt: –43 °C
Density: 1.244 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00000533
CAS Number: 348-51-6
EINECS: 206-476-8
UN: 1993
ADR: 3 III