1-Chloro-3-fluorobenzene 98%
Supplier: Apollo Scientific
Danger
PC1850-100G PC1850-25G PC1850-500G
APOSPC1850-100GEA 77 EUR
APOSPC1850-100G APOSPC1850-25G APOSPC1850-500G
1-Chloro-3-fluorobenzene 98%
1-Chloro-3-fluorobenzene
Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 126…128 °C
Density: 1.219 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000569
CAS Number: 625-98-9
EINECS: 210-919-0
UN: 1993
ADR: 3 II