1-Chloro-4-fluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: Chloro-1-fluoro-4-benzène, 4-Chlorofluorobenzene
PC1860-250G PC1860-25G
APOSPC1860-250GEA 139 EUR
APOSPC1860-250G APOSPC1860-25G
1-Chloro-4-fluorobenzene 98%
1-Chloro-4-fluorobenzene
Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 129…130 °C
Melting Pt: –27 °C
Density: 1.226 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00000603
CAS Number: 352-33-0
EINECS: 206-521-1
UN: 1993
ADR: 3 III