2-Chloro-4-fluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
PC1860C-5G PC1860C-1G
APOSPC1860C-5GEA 31.9 EUR
APOSPC1860C-5G APOSPC1860C-1G
2-Chloro-4-fluorobenzenesulphonyl chloride 97%
2-Chloro-4-fluorobenzenesulphonyl chloride
Formula: C₆H₃Cl₂FO₂S
Boiling Pt: 243…244 °C
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052957
CAS Number: 85958-57-2
UN: 3265
ADR: 8,III