2-Chloro-4-fluorobenzoic acid 99%
Supplier: Apollo Scientific
Warning
PC1860L-100G PC1860L-25G PC1860L-500G
APOSPC1860L-100GEA 43.5 EUR
APOSPC1860L-100G APOSPC1860L-25G APOSPC1860L-500G
2-Chloro-4-fluorobenzoic acid 99%
2-Chloro-4-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 181…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00010615
CAS Number: 2252-51-9
EINECS: 218-845-0