2-Chloro-6-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC1861-100G PC1861-1KG PC1861-250G
APOSPC1861-100GEA 30.6 EUR
APOSPC1861-100G APOSPC1861-1KG APOSPC1861-250G
2-Chloro-6-fluorobenzoic acid 97%
2-Chloro-6-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Boiling Pt: 272
Melting Pt: 155…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00002417
CAS Number: 434-75-3
EINECS: 207-105-2