3-Chloro-2-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC1861C-25G PC1861C-100G PC1861C-1G
APOSPC1861C-25GEA 33.5 EUR
APOSPC1861C-25G APOSPC1861C-100G APOSPC1861C-1G
3-Chloro-2-fluorobenzoic acid 98%
3-Chloro-2-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00042506
CAS Number: 161957-55-7