4-Chloro-2-fluorobenzoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC1861G-100G
PC1861G-500G
APOSPC1861G-100GEA
63.8
EUR
APOSPC1861G-100G
APOSPC1861G-500G
4-Chloro-2-fluorobenzoic acid 98%
4-Chloro-2-fluorobenzoic acid
Formula:
C₇H₄ClFO₂ MW: 174.56 g/mol Melting Pt: 203…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042468 CAS Number: 446-30-0 EINECS: 207-162-3 |