3-Chloro-4-fluorobenzotrifluoride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene
PC1863L-25G PC1863L-100G PC1863L-500G
APOSPC1863L-25GEA 31.9 EUR
APOSPC1863L-25G APOSPC1863L-100G APOSPC1863L-500G
3-Chloro-4-fluorobenzotrifluoride 98%
3-Chloro-4-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Boiling Pt: 137 °C
Density: 1.49 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00042207
CAS Number: 78068-85-6
UN: 1993
ADR: 3 III