2-Chloro-6-fluorobenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC1863W-100G PC1863W-25G PC1863W-5G
APOSPC1863W-100GEA 115 EUR
APOSPC1863W-100G APOSPC1863W-25G APOSPC1863W-5G
2-Chloro-6-fluorobenzoyl chloride 97%
2-Chloro-6-fluorobenzoyl chloride
Formula: C₇H₃Cl₂FO
Boiling Pt: 106…108 °C/15mmHg
Density: 1.43 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00178746
CAS Number: 79455-63-3
EINECS: 279-162-1
UN: 3265
ADR: 8,II