2-Chloro-6-fluorobenzyl alcohol 99%
Supplier: Apollo Scientific
Warning
Synonyms: (2-Chloro-6-fluorophenyl)methanol
PC1864-25G PC1864-100G PC1864-500G
APOSPC1864-25GEA 37.7 EUR
APOSPC1864-25G APOSPC1864-100G APOSPC1864-500G
2-Chloro-6-fluorobenzyl alcohol 99%
2-Chloro-6-fluorobenzyl alcohol
Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 232
Melting Pt: 41…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004608
CAS Number: 56456-50-9
EINECS: 260-192-9