2-Chloro-6-fluorobenzylamine 98%
Supplier: Apollo Scientific
Danger
PC1864D-25G PC1864D-10G PC1864D-5G
APOSPC1864D-25GEA 58 EUR
APOSPC1864D-25G APOSPC1864D-10G APOSPC1864D-5G
2-Chloro-6-fluorobenzylamine 98%
2-Chloro-6-fluorobenzylamine
Formula: C₇H₇ClFN
MW: 159.59 g/mol
Boiling Pt: 91…93 °C/15mmHg
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042458
CAS Number: 15205-15-9
EINECS: 239-259-1
UN: 2735
ADR: 8 II