2-(2-Chloro-6-fluorobenzylthio)ethylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Aminoethyl 2-chloro-6-fluorobenzyl sulphide
PC1864Q-1G PC1864Q-5G PC1864Q-250MG
APOSPC1864Q-1GEA 109 EUR
APOSPC1864Q-1G APOSPC1864Q-5G APOSPC1864Q-250MG
2-(2-Chloro-6-fluorobenzylthio)ethylamine 97%
2-(2-Chloro-6-fluorobenzylthio)ethylamine
Formula: C₉H₁₁ClFNS
MW: 219.71 g/mol
Boiling Pt: 140 °C/6mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052683
CAS Number: 175136-76-2
UN: 3261
ADR: 8,III