1-Chloro-1-fluoroethylene

Supplier: Apollo Scientific
Warning

PC1865-1KG PC1865-25G PC1865-5G PC1865-100G
APOSPC1865-1KGEA 0 EUR
APOSPC1865-1KG APOSPC1865-25G APOSPC1865-5G APOSPC1865-100G
1-Chloro-1-fluoroethylene
1-Chloro-1-fluoroethylene

Delivery information: A delivery charge of this item to mainland UK will be applied.

Formula: C₂H₂ClF
MW: 80.49 g/mol
Boiling Pt: –24 °C
Melting Pt: –169 °C
Storage Temperature: Ambient
CAS Number: 2317-91-1
UN: 3161
ADR: 2.1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR