1-Chloro-2-fluoroethylene 97%

Supplier: Apollo Scientific
Danger

Synonyms: E/Z 1-Chloro-2-fluoroethylene, FC1131

PC1866-25G PC1866-5G
APOSPC1866-25GEA 915 EUR
APOSPC1866-25G APOSPC1866-5G
1-Chloro-2-fluoroethylene 97%
1-Chloro-2-fluoroethylene
Formula: C₂H₂ClF
MW: 80.49
Boiling Pt: 10 °C
Storage Temperature: Ambient
CAS Number: 460-16-2
UN: 3161
ADR: 2.1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR