1-Chloro-2-fluoro-5-iodobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-1-fluoro-4-iodobenzene, 3-Chloro-4-fluoroiodobenzene
PC1868L-100G PC1868L-25G
APOSPC1868L-100GEA 255 EUR
APOSPC1868L-100G APOSPC1868L-25G
1-Chloro-2-fluoro-5-iodobenzene 98%
1-Chloro-2-fluoro-5-iodobenzene
Formula: C₆H₃ClFI
Boiling Pt: 94…95 °C/15mmHg
Density: 2.02 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042210
CAS Number: 156150-67-3