1-Chloro-4-fluoro-2-nitrobenzene 99%
Supplier: Apollo Scientific
Warning
PC1869-25G PC1869-5G PC1869-100G
APOSPC1869-25GEA 29 EUR
APOSPC1869-25G APOSPC1869-5G APOSPC1869-100G
1-Chloro-4-fluoro-2-nitrobenzene 99%
1-Chloro-4-fluoro-2-nitrobenzene
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Boiling Pt: 237…239 °C
Melting Pt: 37…40 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00084853
CAS Number: 345-17-5
EINECS: 206-456-9