4-Chloro-2-fluoro-1-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-2-fluoronitrobenzene
PC1875-100G PC1875-25G PC1875-5G
APOSPC1875-100GEA 165 EUR
APOSPC1875-100G APOSPC1875-25G APOSPC1875-5G
4-Chloro-2-fluoro-1-nitrobenzene 98%
4-Chloro-2-fluoro-1-nitrobenzene
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Melting Pt: 48…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042211
CAS Number: 700-37-8
EINECS: 211-842-5