4-Chloro-3-fluorophenol 98%
Supplier: Apollo Scientific
|
Warning
|
PC1880G-25G
PC1880G-5G
PC1880G-100G
APOSPC1880G-25GEA
53.8
EUR
APOSPC1880G-25G
APOSPC1880G-5G
APOSPC1880G-100G
4-Chloro-3-fluorophenol 98%
4-Chloro-3-fluorophenol
Formula:
C₆H₄ClFO MW: 146.55 g/mol Boiling Pt: 84 °C/44mmHg Melting Pt: 50…56 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042583 CAS Number: 348-60-7 |