2-Chloro-4-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4-fluoro-1-methylbenzene
PC1890-250G PC1890-1KG PC1890-5KG PC1890-10KG
APOSPC1890-250GEA 27.7 EUR
APOSPC1890-250G APOSPC1890-1KG APOSPC1890-5KG APOSPC1890-10KG
2-Chloro-4-fluorotoluene 98%
2-Chloro-4-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 154…156 °C
Density: 1.198 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00000572
CAS Number: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: 3 III