3-Chloro-2-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-fluoro-3-methylbenzene
PC1915-100G PC1915-25G PC1915-5G
APOSPC1915-100GEA 261 EUR
APOSPC1915-100G APOSPC1915-25G APOSPC1915-5G
3-Chloro-2-fluorotoluene 98%
3-Chloro-2-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 92…94 °C/86mmHg
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00042115
CAS Number: 85089-31-2
UN: 1993
ADR: 3,III