4-Chloro-2-fluorotoluene 99%
Supplier: Apollo Scientific
Danger
PC1920-25G PC1920-100G PC1920-250G PC1920-1KG PC1920-5G PC1920-500G PC1920-5KG
APOSPC1920-25GEA 24.9 EUR
APOSPC1920-25G APOSPC1920-100G APOSPC1920-250G APOSPC1920-1KG APOSPC1920-5G APOSPC1920-500G APOSPC1920-5KG
4-Chloro-2-fluorotoluene 99%
4-Chloro-2-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 158 °C
Density: 1.186 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000571
CAS Number: 452-75-5
UN: 1993
ADR: 3 III