6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
Synonyms: 6-Chloro-2-(trifluoromethyl)quinolin-4-ol
PC1931F-25G PC1931F-5G
APOSPC1931F-25GEA 1160 EUR
APOSPC1931F-25G APOSPC1931F-5G
6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃NO
MW: 247.61 g/mol
Melting Pt: 280…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00276583
CAS Number: 18706-21-3