3-Chloro-4-iodobenzotrifluoride 97%
Supplier: Apollo Scientific
Warning
PC1931J-100G PC1931J-25G PC1931J-5G
APOSPC1931J-100GEA 198 EUR
APOSPC1931J-100G APOSPC1931J-25G APOSPC1931J-5G
3-Chloro-4-iodobenzotrifluoride 97%
3-Chloro-4-iodobenzotrifluoride
Formula: C₇H₃ClF₃I
MW: 306.45 g/mol
Boiling Pt: 58…60 °C/0.2mmHg
Density: 1.9802 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042562
CAS Number: 141738-80-9